DBCO-PEG5-NHS ester
Lieferant: AAT BIOQUEST
4526
AATB4526EA
153
EUR
AATB4526
DBCO-PEG5-NHS ester
DBCO-PEG5-NHS ester
DBCO-PEG5-NHS Ester is a specific and efficient reagent that reacts with primary amines (e.g., lysine residues or aminosilane-coated surfaces) at pH 7-9 to form a covalent bond.
- Stable for 12 months
The hydrophilic polyethylene glycol (PEG) spacer arm in these reagents imparts water solubility to the labeled molecule, reducing aggregation of labeled proteins stored in solution.
Bitte beachten For Research Use Only