DBCO Sulfo-NHS ester
Lieferant: AAT Bioquest
4527
4528
AATB4527EA
130
EUR
AATB4527
AATB4528
DBCO Sulfo-NHS ester
DBCO-Sulfo-NHS Ester is a water-soluble version of the popular DBCO NHS Ester (#4529).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only