DBCO-NHS ester zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
N-Succinimidyl 4-[(5-Aza-3,4:7,8-dibenzocyclooct-1-yne)-5-yl]-4-oxobutyrate, DBCO-NHS
4529
AATB4529EA
152
EUR
AATB4529
DBCO-NHS ester zur biomolekularen Markierung
DBCO-NHS ester
Cu-free click reaction, i.e., the strain-promoted alkyne-azide cycloaddition (SPAAC) is a bioorthogonal reaction utilizing a pair of reagents, cyclooctynes and azides that exclusively and efficiently react with each other while remain inert to naturally occurring functional groups such as hydroxy, amino and thiol groups.
- Improved crosslinking efficiency
- High water solubility
Formel:
C₂₃H₁₈N₂O₅ Molecular Weight: 402,41 g/mol |
MDL:
MFCD24386367 CAS-Nummer: 1353016-71-3 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...