Biotin Styramide "Superior Replacement for Biotin Tyramide"
Lieferant: AAT BIOQUEST
45300
AATB45300EA 614 EUR
AATB45300
Biotin Styramide "Superior Replacement for Biotin Tyramide"
Biotin Styramide
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive methods that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 to 50 times higher than the widely used tyramide (TSA) reagents.
  • 10 to 50× higher fluorescence
  • Improved precision and sensitivity
  • More efficient use of primary antibody
  • Enhanced water solubility
  • More photostable
Bitte beachten For research use only.