Propargyl-PEG1-NHS ester zur biomolekularen Markierung

Lieferant: AAT BIOQUEST
Achtung

Synonyme: N-Succinimidyl 3-(propargyloxy)propionate, Propargyl-dPEG®1-NHS ester

4531
AATB4531EA 152 EUR
AATB4531
Propargyl-PEG1-NHS ester zur biomolekularen Markierung
Propargyl-PEG1-NHS ester

Alkyne Modifier I, NHS ester (propargyl-NHS ester) is an alkyne-containing reagent that reacts with primary amines at pH 7-9 forming a stale amide bond.


  • Improved crosslinking efficiency
  • High water solubility

Formel: C₁₀H₁₁NO₅
Molecular Weight: 225,2 g/mol
MDL: MFCD13184973
CAS-Nummer: 1174157-65-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bab3f8537e57a4690ddf8b5221c1f5b37e1fde2&vid=66d0b0c6f100b9f7abd78b3d1bc254b799bde76e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR