DNP Styramide "Superior Replacement for DNP tyramide"
Lieferant: AAT BIOQUEST
45310
AATB45310EA
614
EUR
AATB45310
DNP Styramide "Superior Replacement for DNP tyramide"
DNP Styramide
Power Styramide™ Signal Amplification (PSA™) system is one of the most sensitive metH₂O₂ds that can detect extremely low-abundance targets in cells and tissues with improved fluorescence signal 10 to 50 times higher than the widely used tyramide (TSA) reagents.
- 10 to 50X higher fluorescence
- Improved precision and sensitivity
- More efficient use of primary antibody
- Enhanced water solubility
- More photostable
Bitte beachten For research use only.