Propargyl-PEG5-NHS ester zur biomolekularen Markierung

Lieferant: AAT BIOQUEST
Achtung

Synonyme: N-Succinimidyl 4,7,10,13,16-Pentaoxanonadec-18-ynoate, 2,5-Dioxopyrrolidin-1-yl 4,7,10,13,16-pentaoxanonadec-18-yn-1-oate, 4,7,10,13,16-Pentaoxanonadec-18-ynoic acid N-Succinimidyl ester, Alkyne-PEG5-NHS ester

4533
AATB4533EA 152 EUR
AATB4533
Propargyl-PEG5-NHS ester zur biomolekularen Markierung
Propargyl-PEG5-NHS ester

Alkyne Modifier II, NHS ester is an alkyne-containing reagent that reacts with primary amines at pH 7 to 9 forming a stale amide bond.


  • Improved crosslinking efficiency
  • High water solubility

Formel: C₁₈H₂₇NO₉
Molecular Weight: 401,41 g/mol
MDL: MFCD22574798
CAS-Nummer: 1393330-40-9
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR