Biotin-PEG4-tyramide
Lieferant: AAT BIOQUEST
45352
AATB45352EA
143
EUR
AATB45352
Biotin-PEG4-tyramide
Biotin-PEG4-tyramide
Biotin-PEG4-tyramide is a reagent that has been used for tyramide signal amplification (TSA) via catalyzed reporter deposition (CARD).
- Increased fluorescence signal
- Suitable for low-abundance targets
- Compatible with all major fluorescence platforms
Bitte beachten For research use only.