Biotin-PEG4-tyramide
Lieferant: AAT BIOQUEST
45352
AATB45352EA 143 EUR
AATB45352
Biotin-PEG4-tyramide
Biotin-PEG4-tyramide
Biotin-PEG4-tyramide is a reagent that has been used for tyramide signal amplification (TSA) via catalyzed reporter deposition (CARD).
  • Increased fluorescence signal
  • Suitable for low-abundance targets
  • Compatible with all major fluorescence platforms
Bitte beachten For research use only.