Sulphorhodamine B acid chloride ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Lissamine rhodamine B sulphonyl chloride
470
AATB470EA
152
EUR
AATB470
Sulphorhodamine B acid chloride ≥95%
Sulphorhodamine B acid chloride
SRB sulfonyl chloride (also called Lissamine rhodamine sulfonyl chloride™) is highly fluorescent.
Formel:
C₂₇H₂₉ClN₂O₆S₂ Molecular Weight: 577,107 g/mol |
CAS-Nummer:
62796-29-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...