mFluor™ Violet 545-streptavidin conjugate Konjugat
Lieferant: AAT BIOQUEST
47250
AATB47250EA
153
EUR
AATB47250
mFluor™ Violet 545-streptavidin conjugate Konjugat
mFluor™ Violet 545-streptavidin conjugate
The mFluor™ Violet 545-streptavidin conjugate is a tetrameric biotin-binding protein (streptavidin) covalently labeled with mFluor™ Violet 545 fluorescent dyes.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only