mFluor™ Violet 550-streptavidin conjugate Konjugat
Lieferant: AAT BIOQUEST
47252
AATB47252EA
153
EUR
AATB47252
mFluor™ Violet 550-streptavidin conjugate Konjugat
mFluor™ Violet 550-streptavidin conjugate
mFluor™ Violet 550-streptavidin conjugate consists of the tetrameric biotin-binding protein (streptavidin) covalently labeled with mFluor™ Violet 550 fluorescent dyes.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only