mFluor™ Violet 590-streptavidin conjugate Konjugat
Lieferant: AAT BIOQUEST
47260
AATB47260EA 153 EUR
AATB47260
mFluor™ Violet 590-streptavidin conjugate Konjugat
mFluor™ Violet 590-streptavidin conjugate
mFluor™ Violet 590-streptavidin conjugate consists of the tetrameric biotin-binding protein (streptavidin) covalently labeled with mFluor™ Violet 590 fluorescent dyes.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only