mFluor™ Violet 610-streptavidin conjugate Konjugat
Lieferant: AAT BIOQUEST
47272
AATB47272EA
153
EUR
AATB47272
mFluor™ Violet 610-streptavidin conjugate Konjugat
mFluor™ Violet 610-streptavidin conjugate
mFluor™ Violet 610-streptavidin conjugate consists of the tetrameric biotin-binding protein (streptavidin) covalently labeled with mFluor™ Violet 610 fluorescent dyes.
- Stable for 12 months
Quality tested in-house