Texas Red-X, succinimidyl ester einfaches Isomer fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Sulphorhodamine 101-X, succinimidyl ester

474
AATB474EA 460 EUR
AATB474
Texas Red-X, succinimidyl ester einfaches Isomer fluoreszierende Probe
Texas Red-X, succinimidyl ester

The amine-reactive Texas Red®-X, succinimidyl ester is a superior replacement to Texas Red (Texas Red® is the trademark of Invitrogen).


  • Low interference with conjugated molecule
  • High reproducibility
  • Bright signal

Formel: C₄₁H₄₄N₄O₁₀S₂
Molecular Weight: 816.94 g/mol
CAS-Nummer: 216972-99-5
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=503921e4bff905aa9474800ea8d3a580af5f953d&vid=bc8f707bdb23b87c2e77006f276e47e81aa16916&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR