Texas Red-X, succinimidyl ester Gemisch aus Isomeren ≥90%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Sulphorhodamine 101-X, succinimidyl ester
475
AATB475EA
307
EUR
AATB475
Texas Red-X, succinimidyl ester Gemisch aus Isomeren ≥90%
Texas Red-X, succinimidyl ester
The amine-reactive Texas Red®-X, succinimidyl ester is a superior replacement to Texas Red (Texas Red® is the trademark of Invitrogen).
Formel:
C₄₁H₄₄N₄O₁₀S₂ Molecular Weight: 816,94 g/mol |
CAS-Nummer:
216972-99-5 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...