Sulphorhodamine 101 acid chloride ≥85%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Texas Red®, Sulphorhodamine 101 sulphonyl chloride
480
AATB480EA
152
EUR
AATB480
Sulphorhodamine 101 acid chloride ≥85%
Sulphorhodamine 101 acid chloride
Sulforhodamine 101 acid chloride (also called Texas Red®) is the most popular labeling reagent of sulfonyl chloride.
Formel:
C₃₁H₂₉ClN₂O₆S₂ Molecular Weight: 625,151 g/mol |
MDL:
MFCD00012406 CAS-Nummer: 82354-19-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...