Texas Red® Hydrazide, Single Isomer
Lieferant: AAT BIOQUEST
481
AATB481EA 230 EUR
AATB481
Texas Red® Hydrazide, Single Isomer
Tests Zelltests
Texas Red® hydrazide reacts with carbonyl compounds such as aldehyde, ketones and acids to give bright red fluorescent conjugates.
  • Identical spectra to Texas Red
  • High Stability
Bitte beachten For research use only.