Texas Red® Cadaverine, Single Isomer
Lieferant: AAT BIOQUEST
482
AATB482EA
230
EUR
AATB482
Texas Red® Cadaverine, Single Isomer
Tests
Zelltests
Texas Red® cadaverine reacts with carbonyl compounds such as aldehyde, ketones and acids to give bright red fluorescent conjugates.
- Identical spectra to Texas Red
- High Stability
Bitte beachten For research use only.