Texas Red® Cadaverine, Single Isomer
Lieferant: AAT BIOQUEST
482
AATB482EA 230 EUR
AATB482
Texas Red® Cadaverine, Single Isomer
Tests Zelltests
Texas Red® cadaverine reacts with carbonyl compounds such as aldehyde, ketones and acids to give bright red fluorescent conjugates.
  • Identical spectra to Texas Red
  • High Stability
Bitte beachten For research use only.