Texas Red® Maleimide, Single Isomer
Lieferant: AAT BIOQUEST
483
AATB483EA 152 EUR
AATB483
Texas Red® Maleimide, Single Isomer
Tests Zelltests
Texas Red® maleimide is an excellent thiol-reactive fluorescent reagent for labelling thiol-containing biological molecules (such as peptides, proteins and thiol-modified oligonucleotides).
  • Identical spectra to Texas Red
  • High Stability
Bitte beachten For research use only.