Texas Red® Maleimide, Single Isomer
Lieferant: AAT BIOQUEST
483
AATB483EA
152
EUR
AATB483
Texas Red® Maleimide, Single Isomer
Tests
Zelltests
Texas Red® maleimide is an excellent thiol-reactive fluorescent reagent for labelling thiol-containing biological molecules (such as peptides, proteins and thiol-modified oligonucleotides).
- Identical spectra to Texas Red
- High Stability
Bitte beachten For research use only.