6-ROX azide ≥90%
Lieferant: AAT BIOQUEST
494
AATB494EA 230 EUR
AATB494
6-ROX azide ≥90%
6-ROX azide
6-ROX azide is an excellent alkyne-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
  • Bright Fluorescence
  • Click Chemistry Conjugation Reagent
Bitte beachten For research use only.