6-ROX alkyne
Lieferant: AAT BIOQUEST
495
AATB495EA 230 EUR
AATB495
6-ROX alkyne
6-ROX alkyne
6-ROX alkyne is an excellent azide-reactive fluorescent reagent for labeling alkyne-containing biological molecules through the well known click chemistry.
  • Bright Fluorescence
  • Click Chemistry Conjugation Reagent
Bitte beachten For research use only.