Native Horseradish Horseradish peroxidase (from Serum), Ofloxacin
Lieferant: AAT BIOQUEST
50019
AATB50019EA
769
EUR
AATB50019
Native Horseradish Horseradish peroxidase (from Serum), Ofloxacin
Proteine und Peptide
Ofloxacin-HRP conjugate is used for developing an ELISA assay for quantifying ofloxacin.
- High immunogenicity
- Useful for antibody development
- ELISA assay building block
Bitte beachten For research use only.