Native Bovine Bovine Serum Albumin (from Serum), Ofloxacin
Lieferant: AAT BIOQUEST
50020
AATB50020EA 769 EUR
AATB50020
Native Bovine Bovine Serum Albumin (from Serum), Ofloxacin
Proteine und Peptide
Ofloxacin-BSA Conjugate is used for developing anti-ofloxacin antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Bitte beachten For research use only.