Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Ofloxacin
Lieferant: AAT BIOQUEST
50021
AATB50021EA
769
EUR
AATB50021
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Ofloxacin
Proteine und Peptide
Ofloxacin-KLH Conjugate is used for developing anti-ofloxacin antibodies that are typically used in an ELISA assay.
- High immunogenicity
- Useful for antibody development
- ELISA assay building block
Bitte beachten For research use only.