FMOC-Asp(5/6-FAM)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5003
AATB5003EA 614 EUR
AATB5003
FMOC-Asp(5/6-FAM)-OH zur biomolekularen Markierung
FMOC-Asp(5/6-FAM)-OH
FMOC-Asp(5/6-FAM)-OH is a building block for in-sequence peptide Asp labeling by FAM.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c03854a055907205542e313bcfa4c0135f0f440&vid=4b6783cc97e82259a2bd293a6ad685bdbbabee50&prid=web&clid=SAPDAM