FMOC-Asp(5/6-FAM)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5003
AATB5003EA
614
EUR
AATB5003
FMOC-Asp(5/6-FAM)-OH zur biomolekularen Markierung
FMOC-Asp(5/6-FAM)-OH
FMOC-Asp(5/6-FAM)-OH is a building block for in-sequence peptide Asp labeling by FAM.
- FRET building block
- Multiple applications
ADR:
5,II |