Native Horseradish Horseradish peroxidase (from Serum), Florfenicol
Lieferant: AAT BIOQUEST
50034
AATB50034EA 769 EUR
AATB50034
Native Horseradish Horseradish peroxidase (from Serum), Florfenicol
Proteine und Peptide
Florfenicol-HRP conjugate is used for developing an ELISA assay for quantifying florfenicol.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Bitte beachten For research use only.