Native Horseradish Horseradish peroxidase (from Serum), Florfenicol
Lieferant: AAT BIOQUEST
50034
AATB50034EA
769
EUR
AATB50034
Native Horseradish Horseradish peroxidase (from Serum), Florfenicol
Proteine und Peptide
Florfenicol-HRP conjugate is used for developing an ELISA assay for quantifying florfenicol.
- High immunogenicity
- Useful for antibody development
- ELISA assay building block
Bitte beachten For research use only.