Native Bovine Bovine Serum Albumin (from Serum), Florfenicol
Lieferant: AAT BIOQUEST
50035
AATB50035EA
769
EUR
AATB50035
Native Bovine Bovine Serum Albumin (from Serum), Florfenicol
Proteine und Peptide
Florfenicol-BSA conjugate is used for developing anti-florfenicol antibodies that are typically used in an ELISA assay.
- High immunogenicity
- Useful for antibody development
- ELISA assay building block
Bitte beachten For research use only.