Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Florfenicol
Lieferant: AAT BIOQUEST
50036
AATB50036EA 769 EUR
AATB50036
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Florfenicol
Proteine und Peptide
Florfenicol-KLH conjugate is used for developing anti-florfenicol antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Bitte beachten For research use only.