Native Bovine Bovine Serum Albumin (from Serum), Spectinomycin
Lieferant: AAT BIOQUEST
50038
AATB50038EA
769
EUR
AATB50038
Native Bovine Bovine Serum Albumin (from Serum), Spectinomycin
Proteine und Peptide
Spectinomycin-BSA conjugate is used for developing anti-spectinomycin antibodies that are typically used in an ELISA assay.
- High immunogenicity
- Useful for antibody development
- ELISA assay building block
Bitte beachten For research use only.