Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Spectinomycin
Lieferant: AAT BIOQUEST
50039
AATB50039EA 769 EUR
AATB50039
Native Megathura crenulata Keyhole limpet hemocyanin (from Serum), Spectinomycin
Proteine und Peptide
Spectinomycin-KLH conjugate is used for developing anti-spectinomycin antibodies that are typically used in an ELISA assay.
  • High immunogenicity
  • Useful for antibody development
  • ELISA assay building block
Bitte beachten For research use only.