FMOC-Asp(5-FAM)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5004
AATB5004EA 769 EUR
AATB5004
FMOC-Asp(5-FAM)-OH zur biomolekularen Markierung
FMOC-Asp(5-FAM)-OH
FMOC-Asp(5-FAM)-OH is a building block for in-sequence peptide Asp labeling by FAM.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=79c6b7842763adf4b45f48865afdd9f069739e42&vid=ea852dc09cd43716f96233bf69647e2b2380e761&prid=web&clid=SAPDAM