FMOC-Asp(5-FAM)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5004
AATB5004EA
769
EUR
AATB5004
FMOC-Asp(5-FAM)-OH zur biomolekularen Markierung
FMOC-Asp(5-FAM)-OH
FMOC-Asp(5-FAM)-OH is a building block for in-sequence peptide Asp labeling by FAM.
- FRET building block
- Multiple applications
ADR:
5,II |