FMOC-Asp(5/6-TAMRA)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5005
AATB5005EA 769 EUR
AATB5005
FMOC-Asp(5/6-TAMRA)-OH zur biomolekularen Markierung
FMOC-Asp(5/6-TAMRA)-OH
FMOC-Asp(5/6-TAMRA)-OH is a building block for in-sequence peptide Asp labeling by TAMRA.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea4b248d6577b8606a0b46af6db1c45bda1f6cf3&vid=334fecc74f5cbd253edef9816227f1e7b33dd815&prid=web&clid=SAPDAM