FMOC-Asp(5-TAMRA)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5006
AATB5006EA 1090 EUR
AATB5006
FMOC-Asp(5-TAMRA)-OH zur biomolekularen Markierung
FMOC-Asp(5-TAMRA)-OH
FMOC-Asp(5-TAMRA)-OH is a building block for in-sequence peptide Asp labeling by TAMRA.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=069ff39875bf642657e3a25a8e252d2c2793dce5&vid=cba3661ce17f414384f115bd7b9ce5da5d768bb9&prid=web&clid=SAPDAM