FMOC-Asp(5-TAMRA)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5006
AATB5006EA
1090
EUR
AATB5006
FMOC-Asp(5-TAMRA)-OH zur biomolekularen Markierung
FMOC-Asp(5-TAMRA)-OH
FMOC-Asp(5-TAMRA)-OH is a building block for in-sequence peptide Asp labeling by TAMRA.
- FRET building block
- Multiple applications
ADR:
5,II |