FMOC-Asp(TF3)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5007
AATB5007EA 1090 EUR
AATB5007
FMOC-Asp(TF3)-OH zur biomolekularen Markierung
FMOC-Asp(TF3)-OH
FMOC-Asp(TF3)-OH is a building block for in-sequence Asp labeling by TF3.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b49c60d60004674b57454291c94391e8d5982bc&vid=b3954914107d78874bc4fbc7833724c5a338a3b5&prid=web&clid=SAPDAM