FMOC-Asp(TF3)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5007
AATB5007EA
1090
EUR
AATB5007
FMOC-Asp(TF3)-OH zur biomolekularen Markierung
FMOC-Asp(TF3)-OH
FMOC-Asp(TF3)-OH is a building block for in-sequence Asp labeling by TF3.
- FRET building block
- Multiple applications
ADR:
5,II |