7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: AMCA, 2H-1-Benzopyran-3-acetic acid, 7-Amino-4-methylcoumarin-3-acetic acid, Aminomethyl coumarin acetic acid, 2-(7-Amino-4-methyl-2-oxo-2H-chromen-3-yl)acetic acid

501
AATB501EA 152 EUR
AATB501
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥95%
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H)

AMCA is one of the brightest blue fluorescent labeling dyes.

Formel: C₁₂H₁₁NO₄
Molecular Weight: 233,22 g/mol
MDL: MFCD00081051
CAS-Nummer: 106562-32-7
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR