FMOC-Glu(5-FAM)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5013
AATB5013EA
769
EUR
AATB5013
FMOC-Glu(5-FAM)-OH zur biomolekularen Markierung
FMOC-Glu(5-FAM)-OH
FMOC-Glu(5-FAM)-OH is a building block for in-sequence peptide Glu labeling by FAM.
- FRET building block
- Multiple applications
ADR:
5,II |