FMOC-Glu(5-FAM)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5013
AATB5013EA 769 EUR
AATB5013
FMOC-Glu(5-FAM)-OH zur biomolekularen Markierung
FMOC-Glu(5-FAM)-OH
FMOC-Glu(5-FAM)-OH is a building block for in-sequence peptide Glu labeling by FAM.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fa7fe28f6fa8f4444a6ca95464fdccb18446482&vid=6468c970f4e87a478916a898ba24fa35afb84efa&prid=web&clid=SAPDAM