FMOC-Glu(5/6-TAMRA)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5014
AATB5014EA
769
EUR
AATB5014
FMOC-Glu(5/6-TAMRA)-OH zur biomolekularen Markierung
FMOC-Glu(5/6-TAMRA)-OH
FMOC-Glu(5/6-TAMRA)-OH is a building block for in-sequence peptide Glu labeling by TAMRA.
- FRET building block
- Multiple applications
ADR:
5,II |