FMOC-Glu(5-TAMRA)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5015
AATB5015EA 1090 EUR
AATB5015
FMOC-Glu(5-TAMRA)-OH zur biomolekularen Markierung
FMOC-Glu(5-TAMRA)-OH
FMOC-Glu(5-TAMRA)-OH is a building block for in-sequence peptide Glu labeling by TAMRA.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=936d84a703e73e1a105918f1fb9d84ef3f86f0b6&vid=b73c56e20bad5a4b6814e02cbf2ef9c68e619363&prid=web&clid=SAPDAM