FMOC-Lys(5/6-FAM)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5042
AATB5042EA
700
EUR
AATB5042
FMOC-Lys(5/6-FAM)-OH zur biomolekularen Markierung
FMOC-Lys(5/6-FAM)-OH
FMOC-Lys(5/6-FAM)-OH is a building block for in-sequence Lys labeling by FAM.
- FRET building block
- Multiple applications
ADR:
5,II |