FMOC-Lys(5-FAM)-OH ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: N-alpha-Fmoc-N-epsilon-5-FAM-L-lysine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthen]-5-ylcarboxamido)hexanoic acid

5043
AATB5043EA 152 EUR
AATB5043
FMOC-Lys(5-FAM)-OH ≥95%
FMOC-Lys(5-FAM)-OH

FMOC-Lys(5-FAM)-OH is a building block for in-sequence Lys labeling by FAM.

Formel: C₄₂H₃₄N₂O₁₀
Molecular Weight: 726,73 g/mol
MDL: MFCD03787910
CAS-Nummer: 1242933-88-5
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR