FMOC-Lys(5/6-TAMRA)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5044
AATB5044EA 307 EUR
AATB5044
FMOC-Lys(5/6-TAMRA)-OH zur biomolekularen Markierung
FMOC-Lys(5/6-TAMRA)-OH
FMOC-Lys(5/6-TAMRA)-OH is a building block for in-sequence Lys labeling by TAMRA.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd309fbf7f0a953f4787a7eea19c1d776ee5d957&vid=36ec41c54861be5d34e81088991e9924471c1c38&prid=web&clid=SAPDAM