FMOC-Lys(5-TAMRA)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5045
AATB5045EA 460 EUR
AATB5045
FMOC-Lys(5-TAMRA)-OH zur biomolekularen Markierung
FMOC-Lys(5-TAMRA)-OH
FMOC-Lys(5-TAMRA)-OH is a building block for in-sequence Lys labeling by TAMRA.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60082682219b9811c6d565019a550e3707d3216c&vid=9c5a2a3290d2b2894ee69bd0377f051807cf2fbd&prid=web&clid=SAPDAM