7-(Diethylamino)coumarin-3-carboxylic acid
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
7ACC1, 7-(Diethylamino)-2-oxo-2H-chromene-3-carboxylic acid, DEAC, acid
505
AATB505EA
152
EUR
AATB505
7-(Diethylamino)coumarin-3-carboxylic acid
7-(Diethylamino)coumarin-3-carboxylic acid
7-Diethylaminocoumarin-3-carboxylic acid can be used to create blue-fluorescent bioconjugates.
Formel:
C₁₄H₁₅NO₄ Molecular Weight: 261,28 g/mol |
MDL:
MFCD00068721 CAS-Nummer: 50995-74-9 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...