FMOC-Lys(TF3)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
Achtung
5051
AATB5051EA 769 EUR
AATB5051
FMOC-Lys(TF3)-OH zur biomolekularen Markierung
FMOC-Lys(TF3)-OH
FMOC-Lys(TF3)-OH is a building block for in-sequence Lys labeling by TF3.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=084f39f2b5c03d54c43a4902cb34aa0b60aa8e7a&vid=de991bf9a9c53e2bf1800cf0ea730ef6853749f5&prid=web&clid=SAPDAM