FMOC-Lys(TF3)-OH zur biomolekularen Markierung
Lieferant: AAT BIOQUEST
|
Achtung
|
5051
AATB5051EA
769
EUR
AATB5051
FMOC-Lys(TF3)-OH zur biomolekularen Markierung
FMOC-Lys(TF3)-OH
FMOC-Lys(TF3)-OH is a building block for in-sequence Lys labeling by TF3.
- FRET building block
- Multiple applications
ADR:
5,II |