7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
2,5-Dioxopyrrolidin-1-yl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate, DEAC, DNP-X acid, SE
506
AATB506EA
152
EUR
AATB506
7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester ≥95%
7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester
The amine-reactive 7-diethylaminocoumarin-3-carboxylic acid, succinimidyl ester can be used to create blue-fluorescent bioconjugates.
Formel:
C₁₈H₁₈N₂O₆ Molecular Weight: 358,35 g/mol |
MDL:
MFCD00467585 CAS-Nummer: 139346-57-9 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...