Xylazine NHS ester
Lieferant: AAT BIOQUEST
50610
AATB50610EA
1300
EUR
AATB50610
Xylazine NHS ester
Xylazine NHS ester
Xylazine NHS ester is an excellent amino-reactive building block for developing assays and tests for detecting Xylazine.
- Stable for 12 months
Quality tested in-house