Xylazine amine
Lieferant: AAT BIOQUEST
50611
AATB50611EA
1300
EUR
AATB50611
Xylazine amine
Xylazine amine
Xylazine amine is an excellent building block for developing assays and tests for detecting Xylazine.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only