Xylazine-Fluorescein Konjugat
Lieferant: AAT BIOQUEST
50616
AATB50616EA
1300
EUR
AATB50616
Xylazine-Fluorescein Konjugat
Xylazine-Fluorescein
Xylazine-Fluorescein conjugate is an excellent fluorescent probe for developing assays and tests for detecting Xylazine.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only