Xylazine-BSA Konjugat
Lieferant: AAT BIOQUEST
50620
AATB50620EA
1300
EUR
AATB50620
Xylazine-BSA Konjugat
Xylazine-BSA
Xylazine-BSA conjugate is an excellent probe for developing ELISA assays and tests for detecting Xylazine.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only