AMCA Azide ≥95%
Lieferant: AAT BIOQUEST
508
AATB508EA 152 EUR
AATB508
AMCA Azide ≥95%
AMCA Azide
AMCA is one of the brightest blue fluorescent labeling dyes.
  • Bright Fluorescence
  • Click Chemistry Conjugation Reagent
Bitte beachten For research use only.